Learn More
IKA™ Works Stirring Accessory, Flexible Coupling
Product Code. 10035181 Shop All IKA Products
Change view
Click to view available options
Quantity:
1 Each
Type:
FK 1 Flexible Coupling
Unit Size:
Each
Product Code. Quantity Type Unit Size
10035181 1 Each FK 1 Flexible Coupling Each
1 options
Product Code. 10035181 Supplier IKA Supplier No. 0002336000
  • ONLINE EXCLUSIVE PRICE
    2860.00 DKK
    2635.22 DKK / Pakke med 1
    Spare 224.78 DKK
    7% Slukket
På lager
For stirring tasks using glass stirring rods
  • Compensates for any structural variances
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Each
Type FK 1 Flexible Coupling