Få mere at vide
Proteintech IHCeasy POLR2F Ready-To-Use IHC Kit
Shop alle Proteintech produkter
Mængde:
50 prøver
Beskrivelse
KHC1640 is a ready-to-use IHC kit for staining of POLR2F. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Tekniske data
Tekniske data
| Mængde | 50 prøver |
| Form | Væske |