missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy CD16 Ready-To-Use IHC Kit

A ready-to-use IHC kit for the staining of CD16.

Brand:  Proteintech KHC0029-50T

Additional Details : Weight : 0.55700kg Transport : UN number : 3316 Chem class : 9 Pack group : III

Product Code. 17476733

  • 2898.85 DKK / 1 set
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications

Specifications

Human
Store at 2°C to 8°C
50 Reactions
FFPE Tissue
50 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers