Få mere at vide
Novus Biologicals™ IGSF4A/SynCAM1/CADM1 Overexpression Lysate
Artikelnummer. 18297714 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Pakningsstørrelse:
0.1mg
Artikelnummer. Mængde Pakningsstørrelse
18297714 0.1 mg 0.1mg
1 options
Artikelnummer. 18297714 Leverandør Novus Biologicals™ Leverandørnr. NBL108638
 Begrænset vare
Forventet forsendelse: 15-10-2026
The IGSF4A/SynCAM1/CADM1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC215477 and based on accession number NM_14333. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens cell adhesion molecule 1 (SynCAM/CADM1 ), transcript variant 1, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only