Learn More
IGHD Mouse anti-Human, CoraLite 488, Clone: 1D1B12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67538-100UL
Additional Details : Weight : 0.00010kg
Product Code. 17274746
  • 2734.91 DKK / 100µL
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
IgD is the major antigen receptor isotype on the surface of most peripheral B-cells. The function of IgD is to signal the B cells to be activated. The relative molecular mass and half-life of secreted IgD is 185 kDa and 2.8 days, respectively. Secreted IgD is produced as a monomeric antibody with two heavy chains of the delta (δ) class, and two Ig light chains. This antibody detects the heavy chain (55-70 kDa) of IgD and secreted IgD (185 kDa) .
Specifications
Specifications
IGHD
Monoclonal
1000 μg/mL
Mouse
Primary
Human
IgG2a
Immunofluorescence
1D1B12
CoraLite 488
100 μL
3495
Liquid