missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Mouse Isotype Control
660.00 DKK - 1996.00 DKK
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Conjugate | PerCP-Cyanine5.5 |
Form | Liquid |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15391180
|
Affymetrix eBioscience
45-4732-82 |
100 μg |
1996.00 DKK
100µg |
In Stock
Log in to see stock. |
|||||
15381180
|
Affymetrix eBioscience
45-4732-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: This eBMG2b antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBMG2b antibody has been tested by flow cytometric analysis of normal human peripheral blood and mouse splenocytes. Use at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2b kappa | |
PerCP-Cyanine5.5 | |
Mouse | |
IgG2b κ | |
Affinity chromatography |
eBMG2b | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.