Learn More
Mouse anti-Rat IgG1 Secondary Antibody, APC, eBioscience™, Invitrogen™
Mouse Monoclonal Secondary Antibody
798.00 DKK - 1638.00 DKK
Specifications
Antigen | Rat IgG1 |
---|---|
Clone | R1-12D10 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15508106
|
Affymetrix eBioscience
17-4812-80 |
25 μg |
798.00 DKK
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518106
|
Affymetrix eBioscience
17-4812-82 |
100 μg |
1638.00 DKK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The R1-12D10 monoclonal antibody recognizes rat IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG1. Applications Reported: This R1-12D10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R1-12D10 antibody has been tested by flow cytometric analysis of cells stained with a rat IgG1 primary. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG1 Monoclonal antibody specifically detects Rat IgG1 in Rat samples. It is validated for Flow Cytometry.Specifications
Rat IgG1 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
R1-12D10 | |
Flow Cytometry | |
APC | |
Mouse | |
Rat | |
IgG1 | |
Affinity chromatography |
For Research Use Only.