Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), APC, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
Brand: Affymetrix eBioscience 17-4888-82
1149.39 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.25000kg
Description
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis. Isotype control should be used at the same concentration as experimental antibody. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Armenian Hamster IgG Control antibody specifically detects Armenian Hamster IgG in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Armenian Hamster IgG | |
eBio299Arm | |
APC | |
IgG; Immunoglobulin G; ImmunoglobulinG | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Armenian Hamster | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |
For Research Use Only.