Få mere at vide
IgG Antibody (SPM556), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34428APC
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
Tekniske data
| IgG | |
| Monoclonal | |
| APC | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse | |
| 75 kDa | |
| 0.1 mL | |
| Immunology | |
| 3500 | |
| Human | |
| Purified |
| Flow Cytometry | |
| SPM556 | |
| Flow Cytometry | |
| IGHG1 | |
| Purified human Ig Gamma Chain | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Korrektion af produktindhold
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion