missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IgG Antibody (ICO-97), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34647PE
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
Tekniske data
| IgG | |
| Monoclonal | |
| PE | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse | |
| 75 kDa | |
| 0.1 mL | |
| Immunology | |
| 3500 | |
| Human | |
| Purified |
| Flow Cytometry | |
| ICO-97 | |
| Flow Cytometry | |
| IGHG1 | |
| Purified human Ig Gamma Chain | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion