Learn More
Invitrogen™ IGFBP-6 Mouse ELISA Kit
Sandwich ELISA Kit
4455.00 DKK - 19610.00 DKK
Specifications
Accession Number | P47880 |
---|---|
Assay Sensitivity | 6 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485283
|
Invitrogen™
EMIGFBP6 |
96 Tests |
4455.00 DKK
1 set |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17185739
|
Invitrogen™
EMIGFBP6X5 |
5 x 96 Tests |
19610.00 DKK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The Mouse IGF Binding Protein 6 (Ms IGFBP-6) ELISA quantitates Ms IGFBP-6 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IGFBP-6. Principle of the method The Mouse IGFBP-6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IGFBP6 encodes for a 216-amino acid protein with a calculated molecular weight of 22,847. The protein was found to be an autocrine growth inhibitor. Recombinant IGFBP6 was shown to inhibit growth of HaCat cells and other keratinocyte cell lines.Specifications
P47880 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
16012 | |
<12% | |
HRP | |
Development | |
IGFBP-6 | |
1 hr. 20 min. |
6 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
Mouse | |
Colorimetric Microplate Reader | |
IGFBP-6 | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |