missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ IGF1R beta Monoclonal Antibody (194Q13)

Beskrivelse
IGF1R beta Monoclonal Antibody for Western Blot, ICC/IF
IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of alpha- and beta-subunits, which are disulfide-linked in a beta-alpha-alpha-beta configuration in the mature receptor. The alpha-subunit is completely extracellular, while the beta-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.
Tekniske data
Tekniske data
| Antigen | IGF1R beta |
| Applikationer | Western Blot, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 194Q13 |
| Koncentration | 0.5 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 1% BSA and 0.1% sodium azide; pH 7.2 |
| Gene | IGF1R |
| Genadgangsnr. | P08069, P24062, Q60751 |
| Gene Alias | A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?