missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I R/IGF1R Antibody (IGF1R/4667), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14094JF549
Additional Details : Weight : 0.00970kg
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
IGF-I R/IGF1R | |
Monoclonal | |
Janelia Fluor 549 | |
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
Recombinant fragment (around aa550-750) of human IGF-I R/IGF1R protein (exact sequence is proprietary) (Uniprot: P08069) | |
0.1 mL | |
Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3480 | |
Store at 4°C in the dark. | |
IgG2c κ |
Flow Cytometry, Immunofluorescence | |
IGF1R/4667 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |