Få mere at vide
IgA Rabbit anti-Mouse, Clone: RM220, Abnova™
Artikelnummer. 16108839
Skift visning
Mængde:
100 μg
Pakningsstørrelse:
100µg
Artikelnummer. Mængde unitSize
16108839 100 μg 100µg
1 options
Artikelnummer. 16108839 Leverandør Abnova Leverandørnr. MAB12771.100ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Edge
Rabbit monoclonal antibody raised against native mouse IgA.
Tekniske data
Antigen IgA
Applikationer ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Klassifikation Monoclonal
Klon RM220
Konjugeret Unconjugated
Oversigt Rabbit monoclonal antibody raised against native mouse IgA.
Fortynding ELISA (0.005 - 0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5 - 2 ug/mL) Immunohistochemistry (0.5 - 2 ug/mL) Immunoprecipitation Western Blot (0.5 - 2 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias IgA/Igha
Gensymboler Igh-2
Værtsarter Rabbit
Immunogen Native purified IgA from mouse hybridoma.
Oprensningsmetode Protein A affinity purification
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 238447
Testspecificitet This antibody specifically reacts to mouse IgA. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG2c, IgG3, IgM, IgE, or human IgA.
Målarter Mouse
Indhold og opbevaring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG