Få mere at vide
Novus Biologicals™ IFN-alpha I/IFNA17 Overexpression Lysate
Artikelnummer. 18160510 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18160510 0.1 mg
1 options
Artikelnummer. 18160510 Leverandør Novus Biologicals™ Leverandørnr. NBP206533
 Begrænset vare
Forventet forsendelse: 09-10-2026
The IFN-alpha I/IFNA17 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_6791. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens IFNA17, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only