Få mere at vide
Novus Biologicals IFI44 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals H00010561-B01P
Produktkode 18310389
  • 4540.00 DKK / 0.05mg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
IFI44 Polyclonal antibody specifically detects IFI44 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
IFI44
Polyclonal
Western Blot 1:500
NP_006408.2
Mouse
Protein A purified
RUO
10561
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.4)
interferon-induced protein 44, interferon-induced, hepatitis C-associated microtubular aggregate protein(44kD), MTAP44p44Microtubule-associated protein 44, P44
IFI44 (NP_006408.2, 1 a.a. - 444 a.a.) full-length human protein. MAVTTRLTRLHEKILQNHFGGKRLSLLYKGSVHGFRNGVLLDRCCNQGPTLTVIYSEDHIIGAYAEESYQEGKYASIILFALQDTKISEWKLGLCTPETLFCCDVTKYNSPTNFQIDGRNRKVIMDLKTMENLGLAQNCTISIQDYEVFRCEDSLDERKIKGVIELRKSLLSALRTYEPYGSLVQQIRILLLGPIGAGKSSFFNSVRSVFQGHVTHQALVGTNTTGISEKYRTYSIRDGKDGKYLPFILCDSLGLSEKEGGLCRDDIFYILNGNIRDRYQFNPMESIKLNHHDYIDSPSLKDRIHCVAFVFDASSIQYFSSQMIVKIKRIRRELVNAGVVHVALLTHVDSMDLITKGDLIEIERCEPVRSKLEEVQRKLGFALSDISVVSNYSSEWELDPVKDVLILSALRRMLWAADDFLEDLPFEQIGNLREEIINCAQGKK
0.05 mg
Primary
Human
Purified