missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ IFI30 Monoclonal Antibody (01)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IFI30.
The protein encoded by this gene is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.
Tekniske data
| IFI30 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P13284 | |
| IFI30 | |
| Recombinant Human IFI30 Protein (Met1-Lys243). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 01 | |
| Unconjugated | |
| IFI30 | |
| Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain | |
| Mouse | |
| Protein A | |
| RUO | |
| 10437 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion