Få mere at vide
Novus Biologicals™ IF3EI Overexpression Lysate
Artikelnummer. 18273294 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18273294 0.1 mg
1 options
Artikelnummer. 18273294 Leverandør Novus Biologicals™ Leverandørnr. NBL110193
 Begrænset vare
Forventet forsendelse: 09-10-2026
The IF3EI Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC2669 and based on accession number NM_1691. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens eukaryotic translation initiation factor 3, subunit L (EIF3L), mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only