missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IDE 1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Induces definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 125 nM). Thought to activate the TGF-β signaling pathway; induces Smad2 phosphorylation and increases levels of Nodal expression.
This compound is mixture of E/Z isomers.
Greener Choice Claims
- Environmental benefits include:
- Ozone-Safe or Safe for the Ozone Layer

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 306.31 |
Purity | >98% |
Molecular Weight (g/mol) | 306.318 |
Chemical Name or Material | IDE 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ IDE 1 >
Spot an opportunity for improvement?Share a Content Correction