Få mere at vide
Ibuprofen-d3 Alcohol, TRC
Artikelnummer. 30179385
Skift visning
Mængde:
1 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179385 1 mg
30179242 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179385 Leverandør Toronto Research Chemicals Leverandørnr. TRCI1400471MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1794979-83-1
Molekylær formel C13H17D3O
Molekylvægt (g/mol) 195.32
Synonym beta-(Methyl-d3)-4-(2-methylpropyl)benzeneethanol, 2-(4-Isobutylphenyl)propanol-d3, 2-(4-Isobutylphenyl)propyl-d3 Alcohol;
IUPAC navn 3,3,3-trideuterio-2-[4-(2-methylpropyl)phenyl]propan-1-ol
SMIL [2H]C([2H])([2H])C(CO)c1ccc(CC(C)C)cc1
Tekniske data
Kemisk navn eller materiale Ibuprofen-d3 Alcohol
Mængde 1 mg
InChI formel InChI=1S/C13H20O/c1-10(2)8-12-4-6-13(7-5-12)11(3)9-14/h4-7,10-11,14H,8-9H2,1-3H3/i3D3
Formel vægt 195.1702