missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibuprofen 2,3-Butylene Glycol Ester, Mikromol™
Description
Ibuprofen 2,3-Butylene Glycol Ester
Specifications
Specifications
| Kemisk navn eller materiale | Ibuprofen 2,3-Butylene Glycol Ester |
| CAS | 95093-59-7 |
| Analyt- eller komponentnavne | Ibuprofen 2,3-Butylene Glycol Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H26 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Ibuprofen 2,3-Butylene Glycol Ester, alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid 2-hydroxy-1-methylpropyl ester |
| InChI formel | InChI=1S/C17H26O3/c1-11(2)10-15-6-8-16(9-7-15)12(3)17(19)20-14(5)13(4)18/h6-9,11-14,18H,10H2,1-5H3 |
| SMIL | CC(C)Cc1ccc(cc1)C(C)C(=O)OC(C)C(C)O |
| IUPAC navn | 3-hydroxybutan-2-yl 2-[4-(2-methylpropyl)phenyl]propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?