Få mere at vide
Ibuprofen-13C,d3, TRC
Artikelnummer. 30403875
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30403875 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403875 Leverandør Toronto Research Chemicals Leverandørnr. TRCI14000425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ibuprofen-13C,d3
CAS 1261394-40-4
Renhedsgrad noter HPLC
Molekylær formel C1213CH15D3O2
Mængde 25 mg
Synonym α-Methyl-13C,d3-4-(isobutyl)phenylacetic Acid, [13C,2H3]-Ibuprofen
InChI formel InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/i3+1D3
SMIL CC(C)CC1=CC=C(C([13C]([2H])([2H])[2H])C(O)=O)C=C1
Molekylvægt (g/mol) 210.29
Procent renhed >95