Få mere at vide
Ibufenac Ethyl Ester, Mikromol™
Beskrivelse
Ibufenac Ethyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ibufenac Ethyl Ester |
| CAS | 15649-02-2 |
| Analyt- eller komponentnavne | Ibufenac Ethyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H20 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 4-(2-Methylpropyl)benzeneacetic acid ethyl ester, Acetic acid, (p-isobutylphenyl)-, ethyl ester (7CI,8CI), Ethyl (4-isobutylphenyl)acetate, Ethyl p-isobutylphenylacetate |
| InChI formel | InChI=1S/C14H20O2/c1-4-16-14(15)10-13-7-5-12(6-8-13)9-11(2)3/h5-8,11H,4,9-10H2,1-3H3 |
| SMIL | CCOC(=O)Cc1ccc(CC(C)C)cc1 |
| IUPAC navn | ethyl 2-[4-(2-methylpropyl)phenyl]acetate |
| Vis mere |