missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibudilast, MedChemExpress
Description
Ibudilast (KC-404
Specifications
Specifications
| Kemisk navn eller materiale | Ibudilast |
| Fysisk form | Solid |
| CAS | 50847-11-5 |
| Farve | White |
| Mængde | 10 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C14H18N2O |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | KC-404 AV-411 MN-166 |
| Opløselighedsinformation | DMSO : ≥ 140 mg/mL (607.88 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?