Få mere at vide
Ibogaine Hydrochloride, LoGiCal
Artikelnummer. 30463138
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463138 10 mg
1 options
Artikelnummer. 30463138 Leverandør LoGiCal Leverandørnr. LGCFOR1901.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ibogaine Hydrochloride
CAS 5934-55-4
Analyt- eller komponentnavne Ibogaine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C20 H26 N2 O . Cl H
Certificeringer/compliance ISO 9001
Synonym Ibogaine Hydrochloride, Ibogamine, 12-methoxy-, hydrochloride (1:1), Ibogaine, hydrochloride (7CI), Ibogaine, monohydrochloride (8CI), Ibogamine, 12-methoxy-, monohydrochloride (9CI), 6,9-Methano-5H-pyrido[1',2':1,2]azepino[4,5-b]indole, ibogamine deriv., NSC 29847
InChI formel InChI=1S/C20H26N2O.ClH/c1-3-13-8-12-9-17-19-15(6-7-22(11-12)20(13)17)16-10-14(23-2)4-5-18(16)21-19;/h4-5,10,12-13,17,20-21H,3,6-9,11H2,1-2H3;1H/t12-,13+,17+,20+;/m1./s1
Løsningstype Neat
SMIL Cl.CC[C@H]1C[C@@H]2C[C@@H]3[C@H]1N(CCc4c3[nH]c5ccc(OC)cc45)C2
IUPAC navn (2R,4S,4aS,12bR)-4-ethyl-2,5-methano-9-methoxy-2,3,4,4a,6,7,12,12b-octahydro-1H-indolo[3,2-d][1]benzazepine monohydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 346.89
Mængde 10 mg
Formel vægt 346.1812 g/mol