Få mere at vide
Ibandronic Acid-d3, TRC
Artikelnummer. 30406131
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30406131 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406131 Leverandør Toronto Research Chemicals Leverandørnr. TRCI12000325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ibandronic Acid-d3
CAS 1130899-41-0
Molekylær formel C9H20D3NO7P2
Mængde 25 mg
Synonym P,P'-[1-Hydroxy-3-[(methyl-d3)pentylamino]propylidene]bisphosphonic Acid, [1-Hydroxy-3-[(methyl-d3)pentylamino]propylidene]bisphosphonic Acid, BPH 24-d3, Ibandronate-d3, [1-Hydroxy-3-[(methyl-d3)pentylamino]propylidene]diphosphonic Acid
InChI formel InChI=1S/C9H23NO7P2/c1-3-4-5-7-10(2)8-6-9(11,18(12,13)14)19(15,16)17/h11H,3-8H2,1-2H3,(H2,12,13,14)(H2,15,16,17)/i2D3
SMIL O=P(O)(O)C(P(O)(O)=O)(O)CCN(C([2H])([2H])[2H])CCCCC
Molekylvægt (g/mol) 322.25