missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ IBA1 Recombinant Rat Monoclonal Antibody (HL1880-RT)
GREENER_CHOICE

Artikelnummer. 18036441
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18036441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18036441 Leverandør Invitrogen™ Leverandørnr. MA550414

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rat Recombinant Monoclonal Antibody

IBA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)

Ionized calcium-binding adapter molecule 1 (IBA1), also known by its gene name AIF1, is a protein expressed predominantly by microglia in the brain and spinal cord. This protein belongs to the EF-hand calcium-binding protein family and plays a crucial role in microglial activation and migration in response to brain injury or neuroinflammation. IBA1's function is integral to microglial motility and phagocytic activity, facilitating the cellular response to pathogenic stimuli and promoting tissue homeostasis and repair in the central nervous system. IBA1 serves as a reliable marker for activated microglia in various neurological disorders, including Alzheimer's disease, Parkinson's disease, and multiple sclerosis, where increased expression correlates with disease progression and severity. The protein's structural features enable it to bind calcium ions, inducing conformational changes that activate signaling pathways essential for microglial function. Its expression is highly regulated by inflammatory cytokines, underpinning its role in neuroimmune responses. Due to its specific expression in microglia during pathological conditions, IBA1 is widely used in research as a marker to study microglial status and activity, and it remains a focal point for understanding microglial involvement in neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen IBA1
Applikationer Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Recombinant Monoclonal
Klon HL1880-RT
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with no preservative
Gene AIF1
Genadgangsnr. O70200, P55008, P55009
Gene Alias AI607846; AIF1; AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; Bart1; BART-1; D17H6S50E; DADB-70P7.8; DASS-82G15.2; Em:AF129756.17; G1; Iba; Iba1; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; ionized calcium-binding adapter molecule 1; IRT1; IRT-1; microglia response factor; Mrf1; MRF-1; protein BART-1; protein G1; testis specific
Gensymboler AIF1
Værtsarter Rat
Immunogen Recombinant fragmemt of human Iba1.
Oprensningsmetode Protein A
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 11629, 199, 29427
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.