missing translation for 'onlineSavingsMsg'
Learn More

IACS-8968 R-enantiomer, MedChemExpress

Product Code. 16454588
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16454588 10 mg 10mg
16441169 1 mg -
16407307 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16454588 Supplier Medchem Express Supplier No. HY112164A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IACS-8968 (R-enantiomer) is the R-enantiomer of IACS-8968. IACS-8968 is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.

IACS-8968 (R-enantiomer) is the R-enantiomer of IACS-8968. IACS-8968 is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.

Specifications

Kemisk navn eller materiale IACS-8968 R-enantiomer
CAS 2239305-67-8
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C17H18F3N5O2
Mængde 10 mg
Synonym IDO/TDO Inhibitor (R-enantiomer)
SMIL O=C(N1)N[C@]2(C(C)(C)CN(C3=CC(C(F)(F)F)=CC4=CN=CN34)CC2)C1=O
Molekylvægt (g/mol) 381.35
Formel vægt 381.35
Procent renhed 98.48%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.