Learn More
Invitrogen™ mirVana™ miRNA Inhibitor
Product Code. 13496696
Change view
Click to view available options
Quantity:
0.25 nmol
1 nmol
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13496696 5 nmol 5nmol
13403089 0.25 nmol Each
13413089 1 nmol Each
13416706 20 nmol 20nmol
13426706 250 nmol 250nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13496696 Supplier Invitrogen™ Supplier No. 4464084
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Forventet forsendelse: 28-12-2026
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications.
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications. Based on the latest version of miRbase, mirVana miRNA Inhibitors are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.
Key features include:
  • High specificity and potency—designed to closely resemble natural miRNAs, ensuring high specificity and potent gene regulation
  • Enhanced stability—chemical modifications enhance stability, making them suitable for long-term studies
  • Broad range of targets—available for a comprehensive list of miRNAs, allowing for extensive research possibilities
  • Optimized for delivery—compatible with various transfection reagents and delivery systems, ensuring efficient uptake by cells
miRNA inhibitors to increase gene expression
mirVana miRNA Inhibitors are molecules designed to specifically bind to and inhibit the activity of miRNAs. By inhibiting miRNAs, mirVana miRNA Inhibitors prevent them from binding to their target mRNAs, thereby allowing the expression of those genes to proceed. This can be useful in research to study gene function, understand disease mechanisms, and develop treatments that modulate gene expression.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C).
Description mirVana™ miRNA Inhibitors, 5nmol, Small, Chemically Modified Single-Stranded RNA, Inhibit Endogenous miRNA, Down-Regulation of miRNA Activity
Quantity 5 nmol