Learn More
BUCHI I-80 Controller for Rotavapor R-80
The I-80 is an easy-to-use interface for conveniently regulating your vacuum manually. Increase control of the evaporation process by setting the pump speed and adjusting the manometer.
Brand:  BUCHI 11I0801
5370.73 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30000638
  • 5675.00 DKK / Each
Only null left
Estimated Shipment: 11-08-2025
Description
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Specifications
Controller Interface
Rotavapor R-80 V-80/180
Communication Cable, Holder
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BUCHI I-80 Controller for Rotavapor R-80 > Controller Interface; For Use With (Equipment): Rotavapor R-80 V-80/180; Includes: Communication Cable, Holder