Learn More
BUCHI I-80 Controller for Rotavapor R-80
The I-80 is an easy-to-use interface for conveniently regulating your vacuum manually. Increase control of the evaporation process by setting the pump speed and adjusting the manometer.
Brand: BUCHI 11I0802
5743.70 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Functional design fulfills essential needs of vacuum regulation
- Saves resources thanks to control of vacuum pump
- High longevity due to robust and durable design
- User-friendly and intuitive handling due to self-explaining user interface
- Easy and accurate vacuum regulation
- Relevant information is displayed at a glance
- High reliability due to centralized control of major process parameters
- Full process control thanks to parameter monitoring
- Proven technology for major applications
Specifications
Controller Interface | |
Rotavapor R-80 V-80/180 | |
Power Pack |