Learn More
BUCHI I-80 Controller for Rotavapor R-80
The I-80 is an easy-to-use interface for conveniently regulating your vacuum manually. Increase control of the evaporation process by setting the pump speed and adjusting the manometer.
Brand:  BUCHI 11I0802
5743.70 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30000615
  • 6070.00 DKK / Each
This item is not returnable. View return policy
Description
Description
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Specifications
Controller Interface
Rotavapor R-80 V-80/180
Power Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BUCHI I-80 Controller for Rotavapor R-80 > Controller Interface; For Use With (Equipment): Rotavapor R-80 V-80/180; Includes: Power Pack