missing translation for 'onlineSavingsMsg'
Learn More

Hypertrehalosemic Hormone (Phormia Terrae-novae), TRC

Product Code. 30392691
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30392691 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392691 Supplier Toronto Research Chemicals Supplier No. TRCH998300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Hypertrehalosemic Hormone (Phormia Terrae-novae)
CAS 129204-82-6
Molekylær formel C47H62N10O13
Mængde 100 mg
Synonym 5-Oxo-L-prolyl-L-leucyl-L-threonyl-L-phenylalanyl-L-seryl-L-prolyl-L-alpha-aspartyl-L-Tryptophanamide, red-pigment-concentrating (Pandalus borealis) 3-L-threonine-7-L-aspartic acid-chromatophorotropin
InChI formel InChI=1S/C47H62N10O13/c1-24(2)18-32(52-41(64)30-15-16-37(60)50-30)44(67)56-39(25(3)59)46(69)54-33(19-26-10-5-4-6-11-26)42(65)55-35(23-58)47(70)57-17-9-14-36(57)45(68)53-34(21-38(61)62)43(66)51-31(40(48)63)20-27-22-49-29-13-8-7-12-28(27)29/h4-8,10-13,22,24-25,30-36,39,49,58-59H,9,14-21,23H2,1-3H3,(H2,48,63)(H,50,60)(H,51,66)(H,52,64)(H,53,68)(H,54,69)(H,55,65)(H,56,67)(H,61,62)/t25-,30-,31?,32+,33+,34?,35+,36+,39+/m1/s1
SMIL CC(C)C[C@H](NC(=O)[C@H]1CCC(=O)N1)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](CO)C(=O)N3CCC[C@H]3C(=O)NC(CC(=O)O)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)N
IUPAC navn 4-[[1-amino-3-(1H-indol-3-yl)-1-oxopropan-2-yl]amino]-3-[[(2S)-1-[(2S)-3-hydroxy-2-[[(2S)-2-[[(2S,3R)-3-hydroxy-2-[[(2S)-4-methyl-2-[[(2R)-5-oxopyrrolidine-2-carbonyl]amino]pentanoyl]amino]butanoyl]amino]-3-phenylpropanoyl]amino]propanoyl]pyrrolidine-2-carbonyl]amino]-4-oxobutanoic acid
Molekylvægt (g/mol) 975.05
Formel vægt 974.4498
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.