Få mere at vide
Thermo Scientific™ Hypersil™ SAS Drop-in Guard Cartridges
Shop alle Thermo Scientific produkter
Diameter (metrisk) Indvendig:
2.1 mm
4.6 mm
Partikelstørrelse:
3 μm
Pakningsstørrelse:
Pakke med 4
Beskrivelse
Kræver
Thermo Scientific UNIGUARD™ direct connection and stand alone holders, not included.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Available in packs of 4
Features of the SAS (C1) Solid Phase
- A short alkyl chain reversed phase material with selectivity for polar and multifunctional compounds
- Least retentive Hypersil phase
- Useful for ion-pair separations
Tekniske data
Tekniske data
| Partikelstørrelse | 3 μm |
| Længde (metrisk) | 10 mm |
| Diameter (metrisk) Indvendig | 4.6 mm |
| Stationær fase | C1 |