Learn More
Thermo Scientific™ HyperSep™ UniGuard Direct-Connect Online SPE Columns
Product Code. 11584785 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Hypercarb
HyperSep Quaternary Amine-Modified Polystyrene DVB
HyperSep Sulfonic Acid-Modified Polystyrene DVB
HyperSep Urea-Modified Polystyrene DVB
Diameter (Metric) Inner:
2.1 mm
3.0 mm
Unit Size:
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Diameter (Metric) Inner Unit Size
11584785 HyperSep Hypercarb 2.1 mm Pack of 4
10259224 HyperSep Urea-Modified Polystyrene DVB 2.1 mm Pack of 4
10187554 HyperSep Urea-Modified Polystyrene DVB 3.0 mm Pack of 4
10259134 HyperSep Sulfonic Acid-Modified Polystyrene DVB 2.1 mm Pack of 4
10783487 HyperSep Sulfonic Acid-Modified Polystyrene DVB 3.0 mm Pack of 4
10005984 HyperSep Quaternary Amine-Modified Polystyrene DVB 2.1 mm Pack of 4
10772147 HyperSep Quaternary Amine-Modified Polystyrene DVB 3.0 mm Pack of 4
10015794 HyperSep Hypercarb 3.0 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11584785 Supplier Thermo Scientific™ Supplier No. 60311501
Forventet forsendelse: 25-11-2026
Clean-up and concentrate target analytes in minutes with these direct-connection SPE cartridges, offering quick, easy connection via a reusable drop-in cartridge holder.
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Column offer quick, easy connection by means of a reusable drop-in guard cartridge holder.
HyperSep UNIGUARD Direct-Connect Online SPE Cartridges attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
TRUSTED_SUSTAINABILITY
Specifications
Description HyperSep Hypercarb; 2.1 mm ID
For Use With (Application) Retention of Highly Polar Compounds
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Hypercarb
Type Online Solid Phase Extraction Cartridge
Quantity Pack of 4
Diameter (Metric) Inner 2.1 mm