Få mere at vide
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL
Shop alle Thermo Scientific produkterBeskrivelse
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
- [bullet] Filter-free design to eliminate dead volume
- [bullet] No contamination from the supporting matrix
- [bullet] Available in many popular chemistries
- [bullet] Mass spectrometry
- [bullet] Desalting
- [bullet] Protein purification
- [bullet] MALDI
- [bullet] Electrophoresis
- [bullet] HPCE
- [bullet] HPLC
- [bullet] CEC
Tekniske data
Tekniske data
| Oversigt | 1 to 10 μL Volume |
| Til brug med (applikation) | Microscale Solid Phase Extraction |
| Til brug med (udstyr) | Micropipettes |
| Produktlinje | HyperSep |
| Sorbent | HyperSep Trypsin |
| Type | SpinTip |
| Volumen (metrisk) | 1 to 10 μL |