Få mere at vide
Thermo Scientific™ HyperSep™ Servo+ Total SPE Cartridges and Well Plates
Artikelnummer. 11392273 Shop alle Thermo Scientific produkter
Oversigt:
1 g 25 ml SPE-søjle 20 stk.
150 mg 6 ml SPE-søjle 30 stk.
200 mg 6 ml SPE-søjle 30 stk.
200 mg/3 ml SPE-søjle 50 stk.
500 mg 6 ml SPE-søjle 30 stk.
500 mg/3 ml SPE-søjle 50 stk.
60 mg 6 ml SPE-søjle 30 stk.
60 mg/3 ml SPE-søjle 50 stk.
96-brønds plade 10 mg 1 pakke
96-brønds plade 30 mg 1 pakke
96-brønds plade 5 mg 1 pakning
Sengens vægt:
5 mg
10 mg
30 mg
60 mg
150 mg
200 mg
500 mg
1 g
Pakningsstørrelse:
Hver
Pakke med 20
Pakke med 30
Pakke med 50
Volumen (metrisk):
3 ml
6 ml
25 ml
Artikelnummer. 11392273
Brand: Thermo Scientific™ 60110103P
Screening for unknown components when analyzing drugs of abuse? These SPE cartridges and plates offer a robust, ready-to-use solution.
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo+ products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo+ Total Cartridges and Well Plates offer a robust, ready-to-use solution for screening of a wide range of unknown components.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Tekniske data
Oversigt 96-brønds plade 30 mg 1 pakke
Sengens vægt 30 mg
Til brug med (applikation) Analysis of Drugs of Abuse
Produktlinje HyperSep
Sorbent HyperSep Servo+ Total
Type 96-Well Plate