Learn More
Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate
Product Code. 13496976 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
25 mg
50 mg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
13496976 25 mg Each
13406986 50 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13496976 Supplier Thermo Scientific™ Supplier No. 60306502
 Restricted Item
Forventet forsendelse: 21-12-2026
Unique material for retention of highly polar compounds.
Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents
  • pH stable 0 to 14
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • Interaction mechanism with polar molecules
TRUSTED_SUSTAINABILITY
Specifications
Bed Weight 25 mg
For Use With (Equipment) Universal Vacuum Manifold
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HypeSep Hypercarb
Type 96-well Plate
Quantity 1/Ea