Få mere at vide
Hyoscyamine N-Oxide Hydrochloride, Mikromol™
Beskrivelse
Hyoscyamine N-Oxide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hyoscyamine N-Oxide Hydrochloride |
| CAS | 200721-88-6 |
| Analyt- eller komponentnavne | Hyoscyamine N-Oxide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H23 N O4 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzeneacetic acid, alpha-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), (alphaS)-, Benzeneacetic acid, alpha-(hydroxymethyl)-, 8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride, [1R,3(S),5S]-[partial]- (9CI) |
| InChI formel | InChI=1S/C17H23NO4.ClH/c1-18(21)13-7-8-14(18)10-15(9-13)22-17(20)16(11-19)12-5-3-2-4-6-12;/h2-6,13-16,19H,7-11H2,1H3;1H/t13-,14+,15+,16-,18?;/m1./s1 |
| SMIL | Cl.C[N+]1([O-])[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](CO)c3ccccc3 |
| IUPAC navn | [(1S,5R)-8-methyl-8-oxido-8-azoniabicyclo[3.2.1]octan-3-yl] (2S)-3-hydroxy-2-phenylpropanoate;hydrochloride |
| Vis mere |