Få mere at vide
Hyoscine N-Oxide Hydrobromide, Mikromol™
Beskrivelse
Hyoscine N-Oxide Hydrobromide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hyoscine N-Oxide Hydrobromide |
| CAS | 6106-81-6 |
| Analyt- eller komponentnavne | Hyoscine N-Oxide Hydrobromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H21 N O5 . Br H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Hyoscine N-Oxide Hydrobromide |
| InChI formel | InChI=1S/C17H21NO5.BrH/c1-18(21)13-7-11(8-14(18)16-15(13)23-16)22-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;1H/t11-,12-,13-,14+,15-,16+,18?;/m1./s1 |
| SMIL | Br.C[N+]1([O-])[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4 |
| IUPAC navn | (αS)-α-(Hydroxymethyl)benzeneacetic acid (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-oxido-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide |
| Vis mere |