Få mere at vide
Hygiena PRO-Clean™ Rapid Protein Residue Test
Artikelnummer. 12789336
Encompass
Skift visning
Pakningsstørrelse:
Pakke med 100
Artikelnummer. Pakningsstørrelse
12789336 Pakke med 100
1 options
Artikelnummer. 12789336 Leverandør Hygiena Leverandørnr. 14037778
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
missing translation for 'validMainframeMsg'
Accurately monitor the cleanliness of food equipment surfaces to help ensure food safety and product quality. Simply swab a surface, release the reagent and if food residue is present the reagent will turn purple. Quickly validates hygiene of surface, allowing immediate corrective action to be taken.
  • Color change provides semi-quantitative measure of surface cleanliness
  • The more contamination present, the quicker the color change to purple and the darker the color
  • PRO-Clean detects protein residues left on a surface after cleaning
  • Results in under 10 minutes
  • No instrumentation required - simple color-changing swab device
  • Detects 80μg protein after 1 minute
  • Detects 50μg protein after 5 minutes
  • Detects 20μg protein after 10 minutes
  • Results color chart printed on label for easy interpretation
  • Patented Snap-Valve™ technology and premoistened swab bud
  • Write-on swab tube label
  • Quick and easy way to monitor HACCP/SSOP program
  • Instant results allow for immediate corrective action
  • No technical training required
Meat processors, Food processors, Restaurants and food services, Grocery stores
Tekniske data
Opbevaringskrav 2°C to 25°C