Få mere at vide
Ansell HyFlex™ 11591 Medium Duty Touchscreen Compatible Cut-resistant Gloves
Artikelnummer. 30279126
Skift visning
Størrelse:
9
10
11
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Størrelse
30279126 9
30279127 10
30279132 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30279126 Leverandør Ansell Leverandørnr. 11591090
  • ONLINE EXCLUSIVE PRICE
    3818.10 DKK
    3511.50 DKK / 12 par
    Spare 306.60 DKK
    8% Slukket
Forventet forsendelse: 10-09-2026
Læg i indkøbskurv
Læg i indkøbskurv
HyFlex™ 11591 Medium Duty Touchscreen Gloves are cut-resistant and have a nitrile palm coating, making them ideal for industrial applications.
  • Works with most touchscreen devices
  • Comfortable, breathable, seamless knitted liner
  • Latex-, silicone-, DMF- and fiber glass-free
  • Reinforced thumb crotch
  • Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting wear
  • 13 Gauge
  • Made of HPPE, Nylon, Spandex, Steel, Graphene and Tungsten
TRUSTED_SUSTAINABILITY
Tekniske data
Belægningsmateriale Nitrile
Farve Black
Bedømmelse af skæremodstand ANSI A9
Hånd Specific
Latex fri Yes
Materiale Nylon
Produkttype Cut- and Abrasion-resistant Glove
Størrelse 9