Få mere at vide
Ansell HyFlex™ 11581 Light Duty Touchscreen Compatible Cut-resistant Gloves
Artikelnummer. 30282354 Shop alle Ansell produkter
Skift visning
Størrelse:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Størrelse
30282354 12
30282352 6
30282353 7
30282350 8
30282349 9
30282348 10
30282351 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Artikelnummer. 30282354 Leverandør Ansell Leverandørnr. 11581120
Forventet forsendelse: 14-09-2026
Læg i indkøbskurv
Læg i indkøbskurv
HyFlex™ 11581 Light Duty Touchscreen Gloves are cut-resistant with a knit wrist cuff, making them ideal for industrial applications.
  • High cut protection with an immediate blade blunting effect outperforming cut protection of gloves in the same category
  • High dexterity and tactility
  • 21-gauge construction makes it the thinnest and lightest glove in its category
  • Touchscreen compatible, so it's ready to use in today's workplaces
  • Extended Use-FORTIX™ technology provides up to 20% more abrasion resistance for longer lasting handling in abrasive conditions
  • Reinforced thumb crotch
  • Made of tungsten, HPPE and nylon
  • Spandex seamless liner
  • DGX nitrile palm coating
TRUSTED_SUSTAINABILITY
Tekniske data
Belægningsmateriale Nitrile
Farve Gray, Black
Bedømmelse af skæremodstand ANSI A6
Hånd Specific
Latex fri Yes
Materiale Nylon
Produkttype Cut-resistant Gloves
Størrelse 12