Få mere at vide
Ansell HyFlex™ 11-949 Microfinished Nitrile-Coated Gloves with Intercept Technology™
Shop alle Ansell produkter
Størrelse:
7
8
9
10
Pakningsstørrelse:
12 par
Beskrivelse
- Offers cut, abrasion and oil resistance in various dip geometries
- Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
- Fortix™ Technology increases abrasion resistance
- Microfinished nitrile coating enhances grip capabilities
- Coating is resistant to residual oils and greases present on handled parts
Tekniske data
Tekniske data
| ANSI Abrasion Rating | ANSI 4 |
| Certificeringer/overholdelse (Europa) | EN 420:2003 + A1:2009, REACH-kompatibel |
| Belægningsmateriale | Nitril |
| Farve | Blå |
| Bedømmelse af skæremodstand | ANSI A2 |
| Hånd | Bestemt |
| Materiale | Opsnappe teknologigarn |
| Størrelse | 8 |
| EN388 Standard | 4242B |
| Til brug med (applikation) | Maintenance, Automotive Assembly, Metal Fabrication, and Glass Manufacturing |
| Vis mere |