Få mere at vide
Ansell Edmont™ HyFlex™ 11-625 Series White Polyurethane Medium Weight Gloves
Shop alle Ansell produkter
Størrelse:
6
7
8
9
10
Pakningsstørrelse:
12 par
Beskrivelse
- Perfect balance of comfort and sure handling with superior cut resistance.
- The liners include an innovative yarn structure that contains an optimal rate of Dyneema™.
- Offers high levels of cut resistance
- The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
- Designed for use in dry, as well as slightly oily environments.
Tekniske data
Tekniske data
| Materiale bagpå | Polyurethane |
| Certificeringer/compliance | CE Mark, Standard CATEGORY II EN 388 : 4342 |
| Manchet materiale | Polyurethane |
| Produkttype | Medium Weight Gloves |
| Størrelse | 9 |
| Handske manchet stil | Knitwrist |
| Liner materiale | Dyneema™ |
| Foring | Spandex/Dyneema™ |
| Palm Type | Coated-palm |
| Mængde | 12 Pairs |
| Vis mere |