Få mere at vide
Ansell™ HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves
Shop alle Ansell produkter
Størrelse:
6
7
8
9
10
11
Pakningsstørrelse:
12 par
Beskrivelse
- Designed for a wide range of lightweight applications in dry or slightly oily environments
- Ideal for workers who need a higher level of fit, touch and precision than that offered by their current gloves
- Also ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries
- Ideal for final assembly, airco, electrical engines, wire harness assembly, adjusters work
Tekniske data
Tekniske data
| Materiale bagpå | Polyurethane |
| Certificeringer/compliance | CE Mark, EN 388 : 2121 |
| Manchet materiale | Polyurethane |
| Belægningsmateriale | Polyurethane |
| Produkttype | Lightweight Gloves |
| Størrelse | 9 |
| Stil | 11-619 |
| Handske manchet stil | Knit |
| Foring | Nylon |
| Palm Type | Coated-palm |
| Vis mere |