Description
- Dark liner and glove mask dirt and reduce glove waste
- Light duty 18 gauge liner
- Ideal for the automotive and metal fabrication industries
Specifications
Specifications
| Back Material | Polyurethane |
| Certifications/Compliance | CE Mark, EN 388 : 2121 |
| Cuff Material | Polyurethane |
| Coating Material | Polyurethane |
| Color | Blue |
| Exterior Finish | No Texture |
| Hand | Specific |
| Latex Free | Yes |
| Material | Nylon |
| Product Type | Lightweight Gloves |
| Show More |
Product Title
¿Detecta una oportunidad de mejora?