Få mere at vide
Ansell HyFlex™ 11-591 Medium Duty Touchscreen Compatible Cut Resistant Gloves
Beskrivelse
- Works with most touchscreen devices
- Comfortable, breathable, seamless knitted liner
- Latex-, silicone-, DMF- and fiber glass-free
- Reinforced thumb crotch
- Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting wear
- 13 Gauge
- Made of HPPE, Nylon, Spandex, Steel, Graphene and Tungsten
Tekniske data
Tekniske data
| Belægningsmateriale | Nitrile |
| Farve | Black |
| Bedømmelse af skæremodstand | Level F |
| Hånd | Specific |
| Latex fri | Yes |
| Materiale | Nylon |
| Produkttype | Cut- and Abrasion-resistant Glove |
| Størrelse | 12 |