Få mere at vide
Ansell HyFlex™ 11-591 Medium Duty Touchscreen Compatible Cut Resistant Gloves
Artikelnummer. 30279128
Skift visning
Størrelse:
12
Artikelnummer. Størrelse
30279128 12
1 options
Artikelnummer. 30279128 Leverandør Ansell Leverandørnr. 11591120
Forventet forsendelse: 29-10-2026
Medium duty touchscreen gloves are cut-resistant and have a nitrile palm coating, making them ideal for industrial applications.
  • Works with most touchscreen devices
  • Comfortable, breathable, seamless knitted liner
  • Latex-, silicone-, DMF- and fiber glass-free
  • Reinforced thumb crotch
  • Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting wear
  • 13 Gauge
  • Made of HPPE, Nylon, Spandex, Steel, Graphene and Tungsten
TRUSTED_SUSTAINABILITY
Tekniske data
Belægningsmateriale Nitrile
Farve Black
Bedømmelse af skæremodstand Level F
Hånd Specific
Latex fri Yes
Materiale Nylon
Produkttype Cut- and Abrasion-resistant Glove
Størrelse 12