missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Hyflex™ 11-427 Series Medium Duty 3/4 Dipped Nitrile Polyurethane Gloves

Catalog Number p-7113233
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides good comfort and dexterity. Ansell Hyflex™ 11-427 Series Medium Duty 3/4 Dipped Nitrile Polyurethane Gloves are lint free and seamless knitted.

  • Mechanical protection gloves
  • Seamless knitted glove, gauge 13
  • Palm fit coating
  • Water based process environmentally friendly innovative coating
  • Silicone-free
  • Good abrasion and blade cut resistance
  • Heat resistant up to 100°C
  • Offers an indicator button (orange) which belong to a range of color code corresponding to a cut level : 1 (green), 3 (orange), 5 (red)

Applications:

  • Aeronautics
  • Assembling metal parts and components
  • Assembly of white goods
  • Finishing works
  • Glass manufacturing
  • Cutting small dry or lightly oiled parts
  • Maintenance
  • Plastics injection and moulding
Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2006,EN 407:2004
Color Gray, Bright Yellow
Material Nitrile
Size 10
Glove Cuff Style Knit
Model 11-427_10
Series 11-427
Length (Metric) 220 to 260 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'