Få mere at vide
Ansell HyFle™ 11-644 Cut-resistant Gloves
Beskrivelse
- Heightened protection: HyFlex™ 11-644 cut-resistant gloves utilize INTERCEPT™ Cut Resistance Technology to perform according to ANSI/ISEA105-2024 CUT A2 and EN ISO B laceration protection standards
- Enhanced durability: These abrasion-resistant gloves' polyurethane coating remains both maneuverable and hard-wearing over time
- Improved ergonomics: The 11-644 model's ergonomic fit and low palm weight makes it perfectly comfortable for highly precise operations
- Genuine practicality: HyFlex™ 11-644 PPE gloves are also sold in vend pack form, for practical replenishment and re-application in real-world conditions
Tekniske data
Tekniske data
| Belægningsmateriale | Polyurethane |
| Farve | Gray |
| Bedømmelse af skæremodstand | Level B |
| Hånd | Specific |
| Latex fri | No |
| Materiale | HPPE, Nylon, Spandex, Polyurethane |
| Produkttype | Industrial Gloves |
| Størrelse | 12 |