Få mere at vide
Hydroxyzine-d8 Dihydrochloride, TRC
Artikelnummer. 30406499
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30406499 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406499 Leverandør Toronto Research Chemicals Leverandørnr. TRCH99650210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hydroxyzine-d8 Dihydrochloride
CAS 1808202-93-8
Renhedsgrad noter HPLC
Molekylær formel C21 D8 H19 Cl N2 O2 . 2 Cl H
Mængde 10 mg
Synonym 2-[2-[4-[(4-Chlorophenyl)-phenylmethyl]-1-piperazinyl-d8]ethoxy]ethanol Dihydrochloride, Alamon-d8, Atarax-d8, Aterax-d8, Durrax-d8, Orgatrax-d8, Quiess-d8
InChI formel InChI=1S/C21H27ClN2O2.2ClH/c22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25;;/h1-9,21,25H,10-17H2;2*1H/i10D2,11D2,12D2,13D2;;
SMIL Cl.Cl.[2H]C1([2H])N(CCOCCO)C([2H])([2H])C([2H])([2H])N(C(c2ccccc2)c3ccc(Cl)cc3)C1([2H])[2H]
IUPAC navn 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]-2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl]ethoxy]ethanol;dihydrochloride
Molekylvægt (g/mol) 455.876
Formel vægt 454.18
Procent renhed >95